Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III) ≥97%

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)titanium(III), Ti(TMHD)3

22-6000-1G 22-6000-25G 22-6000-5G
100197-098EA 181.17 USD
100197-098 100210-620 100209-246
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III) ≥97%
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')titanium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Ti
Boiling Pt: 75 °C (0.1 mmHg) (subl.)
MDL Number: MFCD00145466
CAS Number: 181418-64-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR