Copper(II) carbonate basic

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Copper (II) carbonate dihydroxide, Copper(II) carbonate, basic, Basic copper carbonate, Copper (II) carbonate-copper (II) hydroxide (1:1)

93-2911-500G 93-2911-2000G
100209-076EA 90.58 USD
100209-076 100197-796
Copper(II) carbonate basic
Copper (II) carbonate basic
Formula: CuCO₃·Cu(OH)₂
Melting Pt: 200 °C (dec.)
Density: 4.0
MDL Number: MFCD00010976
CAS Number: 12069-69-1
EINECS: 235-113-6
UN: 3288
ADR: 6.1,III
Merck Index: 13,02658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR