Calcium carbonate ≥99.95% (Ca basis)

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Marble

93-2011-100G 93-2011-500G
100209-040EA 96.25 USD
100209-040 100210-238
Calcium carbonate ≥99.95% (Ca basis)
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2.930
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR