Phosphoryl trichloride ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Phosphorus (V) oxide chloride, Phosphorus oxychloride, Phosphoryl chloride, Phosphorus(V) oxychloride

93-1543-250G 93-1543-1000G
100209-024EA 94.83 USD
100209-024 100204-674
Phosphoryl trichloride ≥98%
Phosphoryl trichloride

Halides for CVD & ALD, Volatile Precursors for CVD

Formula: POCl₃
Boiling Pt: 105.3 °C
Melting Pt: 2 °C
Density: 1.675
Storage Temperature: Ambient
MDL Number: MFCD00011443
CAS Number: 10025-87-3
EINECS: 233-046-7
UN: 1810
ADR: 6.1,I
Merck Index: 12,07506

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR