Ammonium heptamolybdate tetrahydrate ACS
Supplier: STREM CHEMICALS INC MS
|
Warning
|
93-0245-100G
93-0245-500G
100207-748EA
45.29
USD
100207-748
100208-990
Ammonium heptamolybdate tetrahydrate ACS
Ammonium heptamolybdate tetrahydrate
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O Density: 2.498 Storage Temperature: Ambient |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 EINECS: 234-722-4 Merck Index: 13,00536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...