Lead ≥99.9995%, wire

Supplier: STREM CHEMICALS INC MS
Warning

82-0200-2MT 82-0200-10MT
100205-214EA 239.2 USD
100205-214 100208-974
Lead ≥99.9995%, wire
Lead
Formula: Pb
Boiling Pt: 1620 °C
Melting Pt: 327 °C
Density: 11.288
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
EINECS: 231-100-4
Merck Index: 12,05410

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR