Bis(benzonitrile)palladium(II) dichloride ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bis(benzonitrile)palladium chloride, Dichlorobis(benzonitrile)palladium(II), Bis(benzonitrile)palladium(II) Dichloride

46-0400-1G 46-0400-5G
100208-914EA 133.05 USD
100208-914 100208-916
Bis(benzonitrile)palladium(II) dichloride ≥99%
Bis(benzonitrile)dichloropalladium(II)
Formula: C₁₄H₁₀Cl₂N₂Pd
Melting Pt: 129…130 °C
MDL Number: MFCD00013123
CAS Number: 14220-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR