Copper(II) oxide ≥99.5% (Cu basis)

Supplier: STREM CHEMICALS INC MS
Warning

29-2945-100G 29-2945-500G
100205-088EA 82.09 USD
100205-088 100208-878
Copper(II) oxide ≥99.5% (Cu basis)
Copper (II) oxide
Formula: CuO
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 1362 °C
Density: 6.3…6.49
Storage Temperature: Ambient
MDL Number: MFCD00010979
CAS Number: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 13,02674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR