Nickel(II) bromide, anhydrous ≥99% (Ni basis)

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel dibromide

28-1140-250G 28-1140-50G
100199-232EA 644 USD
100199-232 100208-874
Nickel(II) bromide, anhydrous ≥99% (Ni basis)
Nickel (II) bromide
Formula: Br₂Ni
Melting Pt: 963 °C
Density: 5.098
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 12,06585

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR