tri-Calcium phosphate (∼37% Ca)

Supplier: STREM CHEMICALS INC MS

Synonyms: Calcium phosphate, Tricalcium orthophosphate, penta-Calcium hydroxide triphosphate, Calcium phosphate tribasic

20-2039-250G 20-2039-1000G 20-2039-5000G
100201-518EA 39.63 USD
100201-518 100208-848 100200-888
tri-Calcium phosphate (∼37% Ca)
tri-Calcium phosphate
Formula: Ca₃(PO₄)₂
Density: 3.14 g/cm³ (20 °C)
MDL Number: MFCD00015984
CAS Number: 7758-87-4
EINECS: 231-840-8
Merck Index: 12,01741

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR