Pentafluorobenzene ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

09-5450-5G 09-5450-25G
100197-352EA 70.77 USD
100197-352 100208-780
Pentafluorobenzene ≥99%
Pentafluorobenzene

Organo Fluorine

Formula: C₆HF₅
Boiling Pt: 85 °C
Melting Pt: –48 °C
Density: 1,514
Flash Pt: 57 °F
Storage Temperature: Ambient
MDL Number: MFCD00000286
CAS Number: 363-72-4
EINECS: 206-658-7
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR