Poly(4-vinylpyridine) (cross-linked) (50-60% H2O), beads

Supplier: STREM CHEMICALS INC MS

Synonyms: Poly-4-vinylpyridine

07-1755-50G 07-1755-250G 07-1755-1000G
100207-934EA 148.62 USD
100207-934 100207-932 100208-760
Poly(4-vinylpyridine) (cross-linked) (50-60% H2O), beads
Poly(4-vinylpyridine) (cross-linked)

Ionic Liquids

Formula: [-CH₂CH(C₅H₄N)CH₂-]n
Boiling Pt: 225 °C/ 260 °C (dec. air/ dec. N₂)
Density: 1,15
MDL Number: MFCD00190588
CAS Number: 9017-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR