1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

45-0169-2G 45-0169-500MG 45-0169-100MG
100197-150EA 2516.55 USD
100197-150 100208-666 100208-664
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₂₂H₃₈BF₄P₂Rh
MDL Number: MFCD09038435
CAS Number: 213343-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR