1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S,S)-Et-DUPHOS-Rh, (+)-1,2-BIS((2S,5S)-2,5-DIETHYLPHOSPHOLANO)BENZENE(CYCLOOCTADIENE)RHODIUM(L)TETRAFLUOROBORATE, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

45-0149-500MG 45-0149-2G 45-0149-100MG
100208-634EA 806.77 USD
100208-634 100197-134 100208-632
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate ≥98%
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate

BPE, DUPHOS

Formula: C₃₀H₄₆BF₄P₂Rh
MDL Number: MFCD01862465
CAS Number: 213343-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR