Tin(IV) oxide ≥99.9% (Sn basis)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

93-5026-500G 93-5026-100G
100205-620EA 181.17 USD
100205-620 100208-572
Tin(IV) oxide ≥99.9% (Sn basis)
Tin (IV) oxide
Formula: O₂Sn
Boiling Pt: ∼2500 °C (1013 hPa)
Melting Pt: 1630 °C
Density: 7.0
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0
Merck Index: 14,08778

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR