Vanadium(IV) oxide ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Vanadium dioxide

93-2309-2G 93-2309-50G
100208-526EA 56.62 USD
100208-526 100210-266
Vanadium(IV) oxide ≥99%
Vanadium (IV) oxide
Formula: O₂V
Boiling Pt: 2700 °C (1013 hPa)
Melting Pt: 1967 °C
Density: 4.339
MDL Number: MFCD00049702
CAS Number: 12036-21-4
EINECS: 234-841-1
UN: 3285
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR