Tributyl borate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-0528-500G 93-0528-100G
100208-490EA 230.71 USD
100208-490 100205-260
Tributyl borate ≥98%
Tributyl borate
Formula: C₁₂H₂₇BO₃
Boiling Pt: 230…235 °C
Melting Pt: –70 °C
Density: 0,853
Flash Pt: 200 °F
MDL Number: MFCD00009434
CAS Number: 688-74-4
EINECS: 211-706-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR