Platinum(II) bromide ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

78-1429-1G 78-1429-5G
100208-464EA 247.69 USD
100208-464 100208-466
Platinum(II) bromide ≥98%
Platinum (II) bromide
Formula: Br₂Pt
Melting Pt: 250 °C (dec.)
Density: 6,65
MDL Number: MFCD00011180
CAS Number: 13455-12-4
EINECS: 236-648-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR