Tungsten boride ≥99.5% (W basis)

Supplier: STREM CHEMICALS INC MS

74-2000-50G 74-2000-10G
100198-248EA 226.46 USD
100198-248 100208-460
Tungsten boride ≥99.5% (W basis)
Tungsten boride
Formula: BW
MDL Number: MFCD00049708
CAS Number: 12007-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR