Barium nitrate ≥99.999% (Ba basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

56-7500-10G 56-7500-50G
100208-452EA 62.28 USD
100208-452 100198-224
Barium nitrate ≥99.999% (Ba basis), PURATREM
Barium nitrate
Formula: Ba(NO₃)₂
Melting Pt: 592 °C
Density: 3.230
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1,II
Merck Index: 13,00986

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR