Tris(2-methylphenyl)phosphine ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

15-8050-25G 15-8050-5G
100208-360EA 363.75 USD
100208-360 100208-362
Tris(2-methylphenyl)phosphine ≥99%
Tris(2-methylphenyl)phosphine

Phosphine

Formula: C₂₁H₂₁P
Boiling Pt: 187…192 °C (0,05 mmHg)
Melting Pt: 125…128 °C
MDL Number: MFCD00008514
CAS Number: 6163-58-2
EINECS: 228-193-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR