Trioctylphosphine oxide ≥90%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: TOPO, Tri-n-octylphosphine Oxide, Trioctylphosphinoxide

15-6660-500G 15-6660-100G
100202-368EA 273.17 USD
100202-368 100208-356
Trioctylphosphine oxide ≥90%
Trioctylphosphine oxide

Nanomaterials, Surfactants for Nano Synthesis

Formula: C₂₄H₅₁OP
Boiling Pt: 409 °C (1013 hPa)
Melting Pt: 51…52 °C
Density: 0,88
Flash Pt: >230 °F
MDL Number: MFCD00002083
CAS Number: 78-50-2
EINECS: 201-121-3
UN: 1759
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR