Bis(trimethylsilyl)methane ≥97%

Supplier: STREM CHEMICALS INC MS
Danger

14-1500-5G 14-1500-1G
100208-310EA 264.68 USD
100208-310 100208-308
Bis(trimethylsilyl)methane ≥97%
Bis(trimethylsilyl)methane
Formula: C₇H₂₀Si₂
Boiling Pt: 133…134 °C
Melting Pt: –71 °C
Density: 0,751
Flash Pt: 55 °F
MDL Number: MFCD00008275
CAS Number: 2117-28-4
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR