N,N'-Bis(3-aminopropyl)ethylenediamine ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 1,5,8,12-Tetraazadodecane

07-1961-100G 07-1961-500G 07-1961-25G
100208-274EA 65.11 USD
100208-274 100201-352 100201-350
N,N'-Bis(3-aminopropyl)ethylenediamine ≥95%
N,N'-Bis(3-aminopropyl)ethylenediamine
Formula: C₈H₂₂N₄
Boiling Pt: 118 °C (0.2 mmHg)
Density: 0,952
Flash Pt: >230 °F
MDL Number: MFCD00008210
CAS Number: 10563-26-5
UN: 2927
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR