Ethylenediamine ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane

07-0570-1000G 07-0570-250G
100208-752EA 176.92 USD
100208-752 100208-268
Ethylenediamine ≥99%
Ethylenediamine
Formula: C₂H₈N₂
Boiling Pt: 118 °C
Melting Pt: 8.5 °C
Density: 0.899
Flash Pt: 93 °F
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbd90391008b8df01337e52ed5760383c063baa&vid=9a49702fe7150a0d3cd56afb586972447ecb6f85&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR