(R)-(+)-2,2'-Diamino-1,1'-binaphthalene ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

07-0327-1G 07-0327-250MG
100195-970EA 318.46 USD
100195-970 100208-264
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene ≥99%
(R)-(+)-2,2'-Diamino-1,1'-binaphthalene
Formula: C₂₀H₁₆N₂
Melting Pt: 242…244 °C
MDL Number: MFCD00145204
CAS Number: 18741-85-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR