Bismuth (III) sulfide ≥99.9% (Bi basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-8309-100G 93-8309-25G
100208-246EA 700.62 USD
100208-246 100208-248
Bismuth (III) sulfide ≥99.9% (Bi basis)
Bismuth (III) sulphide
Formula: Bi₂S₃
Melting Pt: 685 °C (dec.)
Density: 7.39
MDL Number: MFCD00014204
CAS Number: 1345-07-9
EINECS: 215-716-0
Merck Index: 12,01329

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR