Lead(II) nitrate ≥99.999% (Pb basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Danger

93-8267-25G 93-8267-100G
100208-242EA 147.2 USD
100208-242 100200-352
Lead(II) nitrate ≥99.999% (Pb basis), PURATREM
Lead (II) nitrate
Formula: Pb(NO₃)₂
Melting Pt: 470 °C (dec.)
Density: 4.53
Storage Temperature: Ambient
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 13,05432

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR