Thallium(I) nitrate ≥99.5% (Tl basis)

Supplier: STREM CHEMICALS INC MS
Danger

93-8109-100G 93-8109-25G
100208-236EA 765.72 USD
100208-236 100208-238
Thallium(I) nitrate ≥99.5% (Tl basis)
Thallium (I) nitrate
Formula: NO₃Tl
Boiling Pt: 430 °C
Melting Pt: 206 °C
Density: 1.273 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011280
CAS Number: 10102-45-1
EINECS: 233-273-1
UN: 2727
ADR: 6.1,II
Merck Index: 12,09400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR