Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95%

Supplier: STREM CHEMICALS INC MS
Warning

93-4217-25G 93-4217-5G
100208-176EA 140.12 USD
100208-176 100202-130
Bis(2,4-pentanedionato)molybdenum(VI) dioxide ≥95%
Bis(2,4-pentanedionato)molybdenum(VI) dioxide
Formula: C₁₀H₁₄MoO₆
Melting Pt: 181 °C
MDL Number: MFCD00011506
CAS Number: 17524-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR