Triethylsilyl trifluoromethanesulfonate ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

14-7910-25G 14-7910-5G
100207-980EA 410.46 USD
100207-980 100196-046
Triethylsilyl trifluoromethanesulfonate ≥99%
Triethylsilyl trifluoromethanesulphonate
Formula: C₇H₁₅F₃O₃SSi
Boiling Pt: 85…86 °C (12 mmHg)
Density: 1.169
Flash Pt: 161 °F
MDL Number: MFCD00000407
CAS Number: 79271-56-0
EINECS: 279-124-4
UN: 1760
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR