1-Butyl-3-methylimidazolium methanesulfonate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: 1-n-Butyl-3-methylimidazolium methanesulfonate, 1-n-Butyl-3-methylimidazolium methanesulphonate

07-0140-5G
100207-930EA 301.48 USD
100207-930
1-Butyl-3-methylimidazolium methanesulfonate ≥98%
1-Butyl-3-methylimidazolium methanesulphonate

Ionic Liquids

Formula: C₉H₁₈N₂O₃S
Density: 1.21
MDL Number: MFCD06798173
CAS Number: 342789-81-5
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR