Ammonium heptamolybdate tetrahydrate ≥99.999% (Mo basis), PURATREM
Supplier: STREM CHEMICALS INC MS
|
Warning
|
02-0750-25G
02-0750-5G
100207-920EA
452.92
USD
100207-920
100201-300
Ammonium heptamolybdate tetrahydrate ≥99.999% (Mo basis), PURATREM
Ammonium heptamolybdate tetrahydrate
Formula:
(NH₄)₆Mo₇O₂₄·4H₂O Melting Pt: 90 °C Density: 2.498 |
MDL Number:
MFCD00167059 CAS Number: 12054-85-2 EINECS: 234-722-4 Merck Index: 13,00536 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...