Indium (III) oxide ≥99.998% (In basis), PURATREM

Supplier: STREM CHEMICALS INC MS

Synonyms: Indium trioxide, Diindium trioxide

93-4906-5G 93-4906-25G
100207-850EA 77.85 USD
100207-850 100197-240
Indium (III) oxide ≥99.998% (In basis), PURATREM
Indium (III) oxide
Formula: In₂O₃
Melting Pt: 715.05 °C
Density: 7.179
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 12,04984

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR