Silver hexafluoroantimonate ≥98%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Silver hexafluoroantimonate(V)

93-4748-5G 93-4748-1G 93-4748-25G
100210-422EA 70.77 USD
100210-422 100207-846 100198-390
Silver hexafluoroantimonate ≥98%
Silver hexafluoroantimonate
Formula: AgF₆Sb
Density: 4.65…4.85 g/cm³ (20 °C)
MDL Number: MFCD00003401
CAS Number: 26042-64-8
EINECS: 247-429-1
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR