Nickel(II) bromide, hydrate

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Nickel dibromide

93-2807-250G 93-2807-50G
100199-278EA 430.28 USD
100199-278 100207-822
Nickel(II) bromide, hydrate
Nickel (II) bromide
Formula: Br₂Ni
Melting Pt: 300 °C (dec.)
Density: 5.09 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011141
CAS Number: 13462-88-9
EINECS: 236-665-0
UN: 3077
ADR: 9,III
Merck Index: 12,06585

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR