Manganese(III) fluoride ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

93-2542-25G 93-2542-5G
100209-052EA 178.34 USD
100209-052 100207-802
Manganese(III) fluoride ≥98%
Manganese (III) fluoride
Formula: F₃Mn
Melting Pt: Dec.
Density: 3.54
MDL Number: MFCD00016223
CAS Number: 7783-53-1
EINECS: 232-006-6
UN: 3288
ADR: 6.1,III
Merck Index: 12,05784

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR