Ammonium heptamolybdate tetrahydrate ACS

Supplier: STREM CHEMICALS INC MS
Warning

93-0245-100G 93-0245-500G
100207-748EA 45.29 USD
100207-748 100208-990
Ammonium heptamolybdate tetrahydrate ACS
Ammonium heptamolybdate tetrahydrate
Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Density: 2.498
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR