Bismuth(III) oxide ≥99.9998% (Bi basis), PURATREM

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: Bismuth Trioxide, Dibismuth trioxide

83-4025-25G 83-4025-100G
100207-746EA 86.34 USD
100207-746 100202-430
Bismuth(III) oxide ≥99.9998% (Bi basis), PURATREM
Bismuth (III) oxide
Formula: BiO₃
Boiling Pt: 1890 °C
Melting Pt: 817 °C
Density: 8.9
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7
Merck Index: 13,01273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR