Trimethylgallium ≥99%

Supplier: STREM CHEMICALS INC MS
Danger

31-2000-100G 31-2000-25G
100207-654EA 6918.4 USD
100207-654 100196-940
Trimethylgallium ≥99%
Trimethylgallium

Metal Alkyls, Volatile Precursors for CVD

Boiling Pt: 55.7 °C
Melting Pt: –15.8 °C
Density: 1,151
Flash Pt: –1 °F
MDL Number: MFCD00014841
CAS Number: 1445-79-0
UN: 3394
ADR: 4.2,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR