1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥99%

Supplier: STREM CHEMICALS INC MS

Synonyms: Dichloro[bis(1,2-diphenylphosphino)ethane]nickel(II)

28-0030-50G 28-0030-2G 28-0030-10G
100207-644EA 568.98 USD
100207-644 100201-564 100207-642
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride ≥99%
1,2-Bis(diphenylphosphino)ethane nickel(II) chloride
Formula: C₂₆H₂₄Cl₂NiP₂
MDL Number: MFCD00013313
CAS Number: 14647-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR