Dicarbonylcyclopentadienylcobalt ≥95%

Supplier: STREM CHEMICALS INC MS
Danger

Synonyms: Cyclopentadienylcobalt dicarbonyl, Dicarbonylcyclopentadienylcobalt(I)

27-0550-2G 27-0550-10G
100207-636EA 73.6 USD
100207-636 100207-634
Dicarbonylcyclopentadienylcobalt ≥95%
Dicarbonylcyclopentadienylcobalt

Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD

Formula: C₇H₅CoO₂
Boiling Pt: 37…38.5 °C (2 mmHg)
Density: 1,35
Flash Pt: 80 °F
MDL Number: MFCD00009589
CAS Number: 12078-25-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR