Methylcyclopentadienylmanganese tricarbonyl ≥97%
Supplier: STREM CHEMICALS INC MS
|
Danger
|
25-1550-5G
25-1550-25G
100207-624EA
111.82
USD
100207-624
100207-622
Methylcyclopentadienylmanganese tricarbonyl ≥97%
Methylcyclopentadienylmanganese tricarbonyl
Metallocenes, Derivatives & Cp Precursors, Volatile Organometallics for CVD & ALD, Volatile Precursors for CVD
Formula:
C₉H₇MnO₃ Boiling Pt: 233 °C Melting Pt: –2.2 °C Density: 1.38 Flash Pt: 205 °F |
MDL Number:
MFCD00001426 CAS Number: 12108-13-3 UN: 2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...