Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥99% (99.9%-Sc)

Supplier: STREM CHEMICALS INC MS

Synonyms: Tris(2,2,6,6-tetramethyl-3,5-heptanedionato)scandium(III), Sc(TMHD)3

21-1000-2G 21-1000-500MG
100207-616EA 482.65 USD
100207-616 100203-752
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III) ≥99% (99.9%-Sc)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')scandium(III)

Metal Beta-diketonates, Metal TMHD, Volatile Precursors for CVD

Formula: C₃₃H₅₇O₆Sc
Boiling Pt: 275 °C (dec.)
Melting Pt: 150…152 °C
MDL Number: MFCD00169875
CAS Number: 15492-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR