(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (S)-3,3'-Dibromo-1,1'-bi-2-naphthol

08-0601-100MG 08-0601-500MG
100207-530EA 140.12 USD
100207-530 100207-532
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol ≥98%
(S)-(-)-3,3'-Dibromo-1,1'-bi-2-naphthol
Formula: C₂₀H₁₂Br₂O₂
Melting Pt: 257…262 °C
MDL Number: MFCD03093629
CAS Number: 119707-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR