1,2,3-Trimethylimidazolium methyl sulfate ≥98%

Supplier: STREM CHEMICALS INC MS
Warning

07-2660-5G
100207-526EA 162.77 USD
100207-526
1,2,3-Trimethylimidazolium methyl sulfate ≥98%
1,2,3-Trimethylimidazolium methyl sulphate

Ionic Liquids

Formula: C₇H₁₄N₂O₄S
Melting Pt: 115 °C
MDL Number: MFCD03788934
CAS Number: 65086-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR