Potassium tetrabromoplatinate(II) ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

78-1967-5G 78-1967-1G
100207-444EA 895.94 USD
100207-444 100206-838
Potassium tetrabromoplatinate(II) ≥99%
Potassium tetrabromoplatinate (II)
Formula: Br₄K₂Pt
MDL Number: MFCD00011376
CAS Number: 13826-94-3
EINECS: 237-536-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR