(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: (R,R)-BDPP, (2R,4R)-Pentane-2,4-diylbis(diphenylphosphine)

15-0432-2G 15-0432-500MG
100211-004EA 992.18 USD
100211-004 100207-342
(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane ≥99%
(2R,4R)-(+)-2,4-Bis(diphenylphosphino)pentane

Phosphine

Formula: C₂₉H₃₀P₂
Melting Pt: 78 °C
MDL Number: MFCD00142324
CAS Number: 96183-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR