4-Fluorophenol ≥99%

Supplier: STREM CHEMICALS INC MS
Warning

Synonyms: p-Fluorophenol, 1-Fluoro-4-hydroxybenzene

09-4090-100G 09-4090-25G
100207-338EA 257.6 USD
100207-338 100202-900
4-Fluorophenol ≥99%
4-Fluorophenol

Organo Fluorine

Formula: C₆H₅FO
Boiling Pt: 185 °C
Melting Pt: 48…51 °C
Density: 1,22 g/cm³ (20 °C)
Flash Pt: 155 °F
Storage Temperature: Ambient
MDL Number: MFCD00002316
CAS Number: 371-41-5
EINECS: 206-736-0
UN: 2923
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR