Holmium trichloride hexahydrate ≥99.9% (Ho basis)

Supplier: STREM CHEMICALS INC MS
Warning

93-6722-10G 93-6722-2500MG 93-6722-50G
100203-562EA 66.52 USD
100203-562 100202-262 100207-306
Holmium trichloride hexahydrate ≥99.9% (Ho basis)
Holmium trichloride hexahydrate
Formula: Cl₃Ho·6H₂O
MDL Number: MFCD00149739
CAS Number: 14914-84-2
EINECS: 233-387-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR